siRNA Details
Gene Symbol: Plag1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019969.3 | NP_064353.2 | 6 | 1627 | ||
GenBank mRNA | AF057366.1 | - | 2 | 1044 | ||
AK135759.1 | - | 1 | 55 | |||
AY574219.1 | - | 6 | 1604 | |||
BC139283.1 | - | 6 | 1577 | 8862305 | ||
BC139285.1 | - | 6 | 1577 | 8862307 |

More Information
Other Species Targeted

Gene Ontology Categories:
