siRNA Details
Gene Symbol: Mtor | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020009.2 | NP_064393.2 | 45 | 6375 | ||
GenBank mRNA | AF152838.1 | - | 45 | 6350 | ||
AK143381.1 | - | 42 | 5786 | |||
AK158715.1 | - | 24 | 2893 | |||
BC112904.1 | - | 45 | 6333 | 6465126 |
