siRNA Details
Gene Symbol: Tdo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019911.2 | NP_064295.2 | 7, 8 | 769 | ||
GenBank mRNA | AB476989.1 | - | 7, 8 | 1243 | ||
AB476990.1 | - | 7, 8 | 1129 | |||
AK132271.1 | - | 7, 8 | 792 | |||
BC018390.1 | - | 7, 8 | 769 | 4159877 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
