siRNA Details
Gene Symbol: Tulp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF105712.1 | - | 13 | 1609 | ||
AK054304.1 | - | 10 | 1587 | |||
AK077200.1 | - | 13 | 1618 | |||
AK077211.1 | - | 10 | 1415 | |||
AK161165.1 | - | 10 | 1566 | |||
BC048740.1 | - | 6 | 748 | |||
BC089545.1 | - | 10 | 1414 | 6770730 | ||
CD422160.1 | - | 3 | 315 | |||
X69827.1 | - | 7 | 871 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
