siRNA Details
Gene Symbol: Tulp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF105712.1 | - | 3, 4 | 126 | ||
AK077200.1 | - | 3, 4 | 135 | |||
BB617192.1 | - | 3, 4 | 135 | |||
BY087813.1 | - | 3, 4 | 168 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
