siRNA Details
Gene Symbol: Alg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019998.3 | NP_064382.3 | 2 | 852 | ||
GenBank mRNA | AB041604.1 | - | 2 | 789 | ||
AB161357.1 | - | 2 | 786 | |||
AK004997.2 | - | 2 | 810 | |||
AK010673.1 | - | 2 | 818 | |||
AK181454.1 | - | 1 | 257 | |||
BC051951.1 | - | 2 | 800 | 5686826 | ||
BC052411.1 | - | 2 | 805 | 5720077 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
