siRNA Details
Gene Symbol: Rec8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020002.3 | NP_064386.2 | 4 | 388 | ||
GenBank mRNA | AK077167.1 | - | 4 | 323 | ||
AK146832.1 | - | 4 | 374 | |||
AU080225.1 | - | 4 | 389 | |||
BC052155.1 | - | 4 | 384 | 6335959 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
