siRNA Details
Gene Symbol: Lat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020044.3 | NP_064428.1 | 11, 12 | 692 | ||
NM_022964.4 | NP_075253.2 | 10, 11 | 656 | |||
GenBank mRNA | AF257136.1 | - | 10, 11 | 636 | ||
AK134721.1 | - | 8, 9 | 384 | |||
AK138953.1 | - | 11, 12 | 651 | |||
AK143533.1 | - | 10, 11 | 657 | |||
AK144261.1 | - | 11, 12 | 631 | |||
AK162321.1 | - | 10, 11 | 615 | |||
AY190024.1 | - | 11, 12 | 657 | |||
BC005804.1 | - | 10, 11 | 585 | 3154073 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
