siRNA Details
Gene Symbol: C1qtnf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204129.1 | NP_001191058.1 | 2, 3 | 279 | ||
NM_001204130.1 | NP_001191059.1 | 2, 3 | 191 | |||
NM_019959.3 | NP_064343.1 | 2, 3 | 269 | |||
GenBank mRNA | AF192499.1 | - | 2, 3 | 217 | ||
AK005484.1 | - | 2, 3 | 238 | |||
AK145104.1 | - | 2, 3 | 192 | |||
BC037636.1 | - | 1, 2 | 184 | |||
BC049894.1 | - | 2, 3 | 220 | 4018549 | ||
CJ056741.1 | - | 2, 3 | 269 | |||
DQ002394.1 | - | 1, 2 | 163 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
