siRNA Details
Gene Symbol: Clec1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204239.1 | NP_001191168.1 | 4 | 754 | ||
NM_001204253.1 | NP_001191182.1 | 3 | 602 | |||
NM_019985.3 | NP_064369.1 | 5 | 850 | |||
GenBank mRNA | AF201457.1 | - | 5 | 561 | ||
AK007925.1 | - | 5 | 830 | |||
AK154961.1 | - | 105 | ||||
BC064054.1 | - | 5 | 850 | 574821 | ||
EF070191.1 | - | 4 | 444 | |||
EF070192.1 | - | 3 | 289 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
