siRNA Details
Gene Symbol: Zbtb33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079513.1 | NP_001072981.1 | 2 | 1438 | ||
NM_020256.2 | NP_064652.2 | 2 | 1612 | |||
GenBank mRNA | AK087417.1 | - | 2 | 1420 | ||
AK089423.1 | - | 2 | 1572 | |||
BC025136.1 | - | 1 | 1900 | |||
BC138647.1 | - | 2 | 1411 | 8861667 | ||
BC138648.1 | - | 2 | 1411 | 8861668 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
