siRNA Details
Gene Symbol: Scamp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301634.1 | NP_001288563.1 | 6, 7 | 804 | ||
NM_001301635.1 | NP_001288564.1 | 5, 6 | 530 | |||
NM_020270.3 | NP_064666.1 | 6, 7 | 687 | |||
GenBank mRNA | AF241833.1 | - | 6, 7 | 773 | ||
BC018613.1 | - | 6, 7 | 651 | 4511417 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
