siRNA Details
Gene Symbol: Ctsr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020284.1 | NP_064680.1 | 7 | 910 | ||
GenBank mRNA | AF245399.1 | - | 7 | 910 | ||
AK005429.1 | - | 7 | 930 | |||
AK014432.1 | - | 7 | 926 | |||
AY014778.1 | - | 7 | 869 | |||
AY943668.1 | - | 7 | 982 | |||
BC100338.1 | - | 7 | 905 | 30712889 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
