siRNA Details
Gene Symbol: Trpm5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020277.2 | NP_064673.2 | 21 | 3089 | ||
GenBank mRNA | AB039952.1 | - | 20 | 3006 | ||
AF228681.1 | - | 21 | 3089 | |||
AJ271092.2 | - | 20 | 3002 | |||
AK029187.1 | - | 1 | 319 | |||
AK035197.1 | - | 17 | 2619 | |||
AK044908.1 | - | 17 | 2704 | |||
AK161030.1 | - | 12 | 1682 | |||
AY280364.1 | - | 21 | 3053 | |||
AY280365.1 | - | 22 | 3139 | |||
BC133712.1 | - | 20 | 3006 | 40143371 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
