siRNA Details
Gene Symbol: Cldn9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020293.3 | NP_064689.2 | 1 | 813 | ||
GenBank mRNA | BC058186.1 | - | 1 | 805 | 5320961 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
