siRNA Details
Gene Symbol: Cldn13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020504.4 | NP_065250.1 | 1 | 412 | ||
GenBank mRNA | AF516681.2 | - | 1 | 408 | ||
AK010869.1 | - | 1 | 408 | |||
BC115481.1 | - | 1 | 328 | 40100580 | ||
BC115482.1 | - | 1 | 328 | 40100581 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
