siRNA Details
Gene Symbol: Tob2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020507.3 | NP_065253.1 | 2 | 789 | ||
GenBank mRNA | AK049766.1 | - | 2 | 736 | ||
AK122537.1 | - | 1 | 1994 | |||
AK143542.1 | - | 2 | 766 | |||
BC011163.1 | - | 1 | 671 | 4194136 | ||
BC021869.1 | - | 2 | 723 | |||
BC094031.1 | - | 2 | 720 | 4945656 |

More Information
Other Species Targeted

Gene Ontology Categories:
