siRNA Details
Gene Symbol: Slc16a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020516.2 | NP_065262.1 | 6 | 1349 | ||
GenBank mRNA | AF019111.2 | - | 6 | 1358 | ||
BC018216.1 | - | 6 | 1349 | 4481698 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
