siRNA Details
Gene Symbol: Lenep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020517.4 | NP_065263.1 | 1 | 132 | ||
GenBank mRNA | AK053703.1 | - | 145 | |||
BC006806.1 | - | 275 | 3152957 | |||
CK627663.1 | - | 1 | 95 | |||
EL956268.1 | - | 1 | 145 |
Gene Symbol: Flad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC009152.1 | - | 1067 |

More Information
Other Species Targeted

Gene Ontology Categories:
