siRNA Details
Gene Symbol: Lenep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020517.4 | NP_065263.1 | 1 | 144 | ||
GenBank mRNA | AK053703.1 | - | 157 | |||
BC006806.1 | - | 287 | 3152957 | |||
CK627663.1 | - | 1 | 107 | |||
EL956268.1 | - | 1 | 157 |
Gene Symbol: Flad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC009152.1 | - | 1079 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
