siRNA Details
Gene Symbol: Icmt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133788.2 | NP_598549.1 | 3 | 464 | ||
GenBank mRNA | AF209926.1 | - | 3 | 246 | ||
AK154135.1 | - | 3 | 396 | |||
AK154410.1 | - | 4 | 487 | |||
BC057132.1 | - | 4 | 483 | |||
BC110695.1 | - | 3 | 451 | 6842587 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
