siRNA Details
Gene Symbol: Srsf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020587.2 | NP_065612.2 | 4 | 639 | ||
GenBank mRNA | AB041587.1 | - | 1 | 77 | ||
AK075769.1 | - | 4 | 607 | |||
AK077648.1 | - | 4 | 625 | |||
AK156989.1 | - | 4 | 574 | |||
BC019437.1 | - | 4 | 599 | 4018178 | ||
BC026944.1 | - | 4 | 1505 | 4016087 | ||
BC040812.1 | - | 4 | 1505 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001108685, BC160850.
Gene Ontology Categories:
