siRNA Details
Gene Symbol: Terf2ip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020584.2 | NP_065609.2 | 3 | 928 | ||
GenBank mRNA | AB041557.1 | - | 3 | 882 | ||
AB041559.1 | - | 3 | 883 | |||
AK041613.1 | - | 3 | 874 | |||
AK051818.1 | - | 3 | 848 | |||
AK081557.1 | - | 3 | 884 | |||
AK084563.1 | - | 3 | 876 | |||
AK087729.1 | - | 3 | 886 | |||
AK148508.1 | - | 3 | 888 | |||
AK148537.1 | - | 3 | 890 | |||
AK161452.1 | - | 3 | 907 | |||
AK165074.1 | - | 3 | 886 | |||
BC012270.1 | - | 3 | 844 | 3483521 | ||
BC017641.1 | - | 3 | 904 | 4483320 |

More Information
Other Species Targeted

Gene Ontology Categories:
