siRNA Details
Gene Symbol: Terf2ip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020584.2 | NP_065609.2 | 1 | 187 | ||
GenBank mRNA | AB041557.1 | - | 1 | 141 | ||
AB041559.1 | - | 1 | 142 | |||
AK041613.1 | - | 1 | 133 | |||
AK047222.1 | - | 1 | 144 | |||
AK051818.1 | - | 1 | 107 | |||
AK053864.1 | - | 1 | 143 | |||
AK081557.1 | - | 1 | 143 | |||
AK084563.1 | - | 1 | 135 | |||
AK087729.1 | - | 1 | 145 | |||
AK148508.1 | - | 1 | 147 | |||
AK148537.1 | - | 1 | 149 | |||
AK161452.1 | - | 1 | 166 | |||
AK163044.1 | - | 1 | 143 | |||
AK165074.1 | - | 1 | 145 | |||
BC012270.1 | - | 1 | 103 | 3483521 | ||
BC017641.1 | - | 1 | 163 | 4483320 | ||
BY129872.1 | - | 1 | 187 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
