siRNA Details
Gene Symbol: Cramp1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020608.3 | NP_065633.2 | 10 | 2003 | ||
GenBank mRNA | AK089393.1 | - | 10 | 2262 | ||
AK129356.1 | - | 1 | 179 | |||
AK147411.1 | - | 10 | 2004 | |||
AK147449.1 | - | 10 | 2004 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
