siRNA Details
Gene Symbol: Srd5a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020611.4 | NP_065636.2 | 6 | 982 | ||
GenBank mRNA | AK040198.1 | - | 6 | 950 | ||
AK075635.1 | - | 6 | 978 | |||
AK085748.1 | - | 6 | 831 | |||
AK139290.1 | - | 6 | 979 | |||
AK162527.1 | - | 6 | 950 | |||
AK164282.1 | - | 2 | 1084 | |||
BC014771.1 | - | 6 | 988 | 4024297 | ||
BC039564.1 | - | 6 | 935 | |||
BC145647.1 | - | 6 | 978 | 40130950 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
