siRNA Details
Gene Symbol: Mogs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020619.2 | NP_065644.2 | 4 | 1321 | ||
GenBank mRNA | AK144946.1 | - | 4 | 1322 | ||
AK150873.1 | - | 1 | 57 | |||
AK151941.1 | - | 1 | 57 | |||
AK152127.1 | - | 1 | 57 | |||
AK157286.1 | - | 4 | 1318 | |||
BC051949.1 | - | 4 | 1321 | 5716189 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
