siRNA Details
Gene Symbol: Psors1c2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020576.2 | NP_065601.2 | 2 | 525 | ||
GenBank mRNA | AF159091.1 | - | 2 | 457 | ||
AF484421.1 | - | 2 | 484 | |||
AK137759.1 | - | 2 | 526 | |||
BC110561.2 | - | 2 | 453 | 40049760 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
