siRNA Details
Gene Symbol: Tmem27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020626.2 | NP_065651.1 | 4, 5 | 411 | ||
GenBank mRNA | AF178085.1 | - | 4, 5 | 411 | ||
AK002337.1 | - | 4, 5 | 357 | |||
AK148534.1 | - | 4, 5 | 408 | |||
BC049912.1 | - | 4, 5 | 333 | 4973401 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
