siRNA Details
Gene Symbol: Gabarapl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020590.4 | NP_065615.1 | 1 | 315 | ||
GenBank mRNA | AB041648.1 | - | 1 | 339 | ||
AF180518.1 | - | 1 | 342 | |||
AK011975.1 | - | 1 | 310 | |||
AK014083.1 | - | 1 | 129 | |||
AK015049.1 | - | 1 | 132 | |||
AK018687.1 | - | 1 | 316 | |||
AK159015.1 | - | 1 | 310 | |||
AK167500.1 | - | 1 | 308 | |||
AK168921.1 | - | 1 | 347 | |||
AK182413.1 | - | 1 | 89 | |||
BC004602.1 | - | 1 | 250 | 3585132 | ||
BC024706.1 | - | 1 | 314 | 3711973 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
