siRNA Details
Gene Symbol: Golga7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042484.1 | NP_001035949.1 | 3 | 327 | ||
NM_020585.2 | NP_065610.2 | 2 | 540 | |||
GenBank mRNA | AB041568.1 | - | 2 | 474 | ||
AK048175.1 | - | 3 | 303 | |||
AK077582.1 | - | 3 | 301 | |||
AK089856.1 | - | 3 | 315 | |||
AK150462.1 | - | 3 | 321 | |||
AK150696.1 | - | 2 | 482 | |||
AK151353.1 | - | 3 | 321 | |||
AK152520.1 | - | 3 | 321 | |||
AK152606.1 | - | 3 | 300 | |||
AK153213.1 | - | 3 | 321 | |||
AK158227.1 | - | 2 | 446 | |||
AK170129.1 | - | 3 | 306 | |||
AK179247.1 | - | 2 | 150 | |||
AK184095.1 | - | 2 | 150 | |||
AK205367.1 | - | 1 | 40 | |||
BC016894.1 | - | 3 | 279 | 4223937 | ||
BC019697.1 | - | 2 | 465 | |||
BC058523.1 | - | 2 | 384 | 5717934 | ||
BC093518.1 | - | 3 | 297 | 3157520 | ||
BC103796.1 | - | 3 | 292 | 6519018 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
