siRNA Details
Gene Symbol: Kcne3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190869.1 | NP_001177798.1 | 3, 4 | 516 | ||
NM_020574.5 | NP_065599.1 | 2, 3 | 215 | |||
GenBank mRNA | AF076532.1 | - | 1, 2 | 116 | ||
AK008744.1 | - | 2, 3 | 204 | |||
AK132773.1 | - | 3, 4 | 508 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
