siRNA Details
Gene Symbol: Stk32c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162540.1 | NP_001156012.1 | 11 | 1134 | ||
NM_021302.3 | NP_067277.2 | 11 | 1434 | |||
GenBank mRNA | AB041542.1 | - | 11 | 1414 | ||
AK046439.1 | - | 11 | 1412 | |||
BC026457.1 | - | 11 | 1419 | 4481218 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
