siRNA Details
Gene Symbol: Noc2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021303.2 | NP_067278.2 | 8 | 880 | ||
GenBank mRNA | AK076287.1 | - | 7 | 922 | ||
AK078560.1 | - | 8 | 856 | |||
AK133834.1 | - | 8 | 880 | |||
AK157765.1 | - | 8 | 873 | |||
AK164071.1 | - | 2 | 2497 | |||
AK184541.1 | - | 4 | 325 | |||
BC020013.1 | - | 8 | 851 | 3663482 |

More Information
Other Species Targeted

Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
