siRNA Details
Gene Symbol: Piwil2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021308.1 | NP_067283.1 | 16 | 2138 | ||
GenBank mRNA | AB032605.1 | - | 16 | 2138 | ||
AF285586.1 | - | 7 | 867 | |||
AK030116.1 | - | 16 | 2082 | |||
AK163647.1 | - | 3 | 393 | |||
BC138444.1 | - | 16 | 2126 | 8861464 | ||
BC145717.1 | - | 16 | 2126 | 40131020 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
