siRNA Details
Gene Symbol: Piwil2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021308.1 | NP_067283.1 | 11 | 1446 | ||
GenBank mRNA | AB032605.1 | - | 11 | 1446 | ||
AF285586.1 | - | 2 | 175 | |||
AK030116.1 | - | 11 | 1390 | |||
BC138444.1 | - | 11 | 1434 | 8861464 | ||
BC145717.1 | - | 11 | 1434 | 40131020 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
