siRNA Details
Gene Symbol: Ntn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021320.3 | NP_067295.2 | 9 | 2115 | ||
GenBank mRNA | AF268066.1 | - | 9 | 1710 | ||
AF281278.1 | - | 9 | 1963 | |||
BC119182.1 | - | 9 | 1797 | 8733931 | ||
BC119184.1 | - | 9 | 1797 | 8733933 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
