siRNA Details
Gene Symbol: Bin3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021328.3 | NP_067303.1 | 8, 9 | 668 | ||
GenBank mRNA | AF244361.1 | - | 8, 9 | 648 | ||
AF271733.1 | - | 8, 9 | 711 | |||
AK005992.1 | - | 8, 9 | 677 | |||
AK040685.1 | - | 8, 9 | 668 | |||
AK170915.1 | - | 8, 9 | 709 | |||
BC026543.1 | - | 8, 9 | 653 | 5040094 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
