siRNA Details
Gene Symbol: Isy1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133934.4 | NP_598695.1 | 6 | 403 | ||
GenBank mRNA | AK017996.1 | - | 6 | 320 | ||
AK173116.1 | - | 371 | ||||
BC020103.1 | - | 6 | 367 | 3673446 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
