siRNA Details
Gene Symbol: Tnfrsf13b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021349.1 | NP_067324.1 | 2, 3 | 351 | ||
GenBank mRNA | AF257673.1 | - | 2, 3 | 351 | ||
AK004668.1 | - | 3, 4 | 428 | |||
BC139392.1 | - | 2, 3 | 408 | 8862414 | ||
BC139393.1 | - | 2, 3 | 408 | 8862415 | ||
BC141867.1 | - | 2, 3 | 351 | 40054144 | ||
BC141868.1 | - | 2, 3 | 351 | 40054146 |
