siRNA Details
Gene Symbol: Sertad2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038625.1 | NP_001033714.1 | 2 | 702 | ||
NM_021372.2 | NP_067347.2 | 2 | 913 | |||
GenBank mRNA | AB041541.1 | - | 2 | 832 | ||
AF366403.1 | - | 1 | 17 | |||
AK076787.1 | - | 2 | 606 | |||
AK132846.1 | - | 2 | 682 | |||
AK132849.1 | - | 2 | 521 | |||
AK142018.1 | - | 2 | 682 | |||
AK146621.1 | - | 2 | 703 | |||
AK146897.1 | - | 2 | 703 | |||
AK147674.1 | - | 3 | 1089 | |||
BC014726.1 | - | 2 | 745 | 4240282 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
