siRNA Details
Gene Symbol: Il20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021380.1 | NP_067355.1 | 2 | 275 | ||
GenBank mRNA | AF224267.1 | - | 2 | 275 | ||
AK078698.1 | - | 2 | 431 | |||
BC111821.1 | - | 2 | 265 | 40049751 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
