siRNA Details
Gene Symbol: Prokr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021381.3 | NP_067356.2 | 2 | 694 | ||
GenBank mRNA | AF236082.1 | - | 1 | 448 | ||
AF487278.1 | - | 1 | 448 | |||
AK134279.1 | - | 2 | 708 | |||
AK156734.1 | - | 2 | 868 | |||
BC059003.1 | - | 2 | 694 | 6848525 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
