siRNA Details
Gene Symbol: Rsad2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021384.4 | NP_067359.2 | 3 | 701 | ||
GenBank mRNA | AF236064.2 | - | 3 | 677 | ||
AF442152.1 | - | 3 | 662 | |||
AK010906.1 | - | 3 | 680 | |||
AK036373.1 | - | 3 | 681 | |||
AK149900.1 | - | 3 | 682 | |||
AK150069.1 | - | 3 | 683 | |||
AK150347.1 | - | 3 | 683 | |||
AK150406.1 | - | 3 | 681 | |||
AK150425.1 | - | 3 | 683 | |||
AK150525.1 | - | 3 | 682 | |||
AK150536.1 | - | 3 | 684 | |||
AK150616.1 | - | 3 | 681 | |||
AK150684.1 | - | 3 | 683 | |||
AK150731.1 | - | 3 | 683 | |||
AK150767.1 | - | 3 | 683 | |||
AK150783.1 | - | 3 | 683 | |||
AK150788.1 | - | 3 | 683 | |||
AK150812.1 | - | 3 | 683 | |||
AK150835.1 | - | 3 | 682 | |||
AK151140.1 | - | 3 | 683 | |||
AK151185.1 | - | 3 | 683 | |||
AK151191.1 | - | 3 | 683 | |||
AK151206.1 | - | 3 | 683 | |||
AK151286.1 | - | 3 | 681 | |||
AK151308.1 | - | 3 | 683 | |||
AK151331.1 | - | 4 | 1118 | |||
AK151339.1 | - | 3 | 681 | |||
AK151389.1 | - | 3 | 683 | |||
AK151410.1 | - | 3 | 682 | |||
AK151412.1 | - | 3 | 681 | |||
AK151414.1 | - | 3 | 682 | |||
AK151476.1 | - | 3 | 683 | |||
AK151502.1 | - | 3 | 683 | |||
AK151538.1 | - | 3 | 683 | |||
AK151767.1 | - | 3 | 683 | |||
AK151770.1 | - | 3 | 681 | |||
AK151825.1 | - | 3 | 681 | |||
AK151833.1 | - | 3 | 681 | |||
AK151846.1 | - | 3 | 681 | |||
AK151906.1 | - | 3 | 683 | |||
AK151909.1 | - | 3 | 681 | |||
AK151940.1 | - | 3 | 681 | |||
AK151954.1 | - | 3 | 683 | |||
AK151971.1 | - | 3 | 683 | |||
AK152050.1 | - | 3 | 681 | |||
AK152065.1 | - | 3 | 683 | |||
AK152075.1 | - | 3 | 683 | |||
AK152235.1 | - | 3 | 681 | |||
AK152239.1 | - | 3 | 683 | |||
AK152241.1 | - | 3 | 683 | |||
AK152310.1 | - | 3 | 681 | |||
AK152354.1 | - | 3 | 681 | |||
AK152493.1 | - | 3 | 683 | |||
AK152600.1 | - | 3 | 681 | |||
AK152601.1 | - | 3 | 683 | |||
AK152631.1 | - | 3 | 681 | |||
AK152644.1 | - | 3 | 683 | |||
AK152664.1 | - | 3 | 683 | |||
AK152701.1 | - | 3 | 681 | |||
AK152796.1 | - | 3 | 681 | |||
AK152861.1 | - | 3 | 681 | |||
AK152880.1 | - | 3 | 681 | |||
AK152895.1 | - | 3 | 683 | |||
AK152934.1 | - | 3 | 683 | |||
AK152962.1 | - | 3 | 681 | |||
AK153003.1 | - | 3 | 683 | |||
AK153024.1 | - | 3 | 683 | |||
AK153062.1 | - | 3 | 685 | |||
AK153077.1 | - | 3 | 681 | |||
AK153078.1 | - | 3 | 681 | |||
AK153232.1 | - | 3 | 683 | |||
AK153234.1 | - | 3 | 683 | |||
AK153306.1 | - | 3 | 681 | |||
AK153318.1 | - | 3 | 683 | |||
AK153324.1 | - | 3 | 683 | |||
AK153350.1 | - | 4 | 1118 | |||
AK153435.1 | - | 3 | 559 | |||
AK153453.1 | - | 3 | 680 | |||
AK153556.1 | - | 3 | 683 | |||
AK159147.1 | - | 3 | 681 | |||
BC057868.1 | - | 3 | 674 | 5327504 | ||
CT010378.1 | - | 3 | 668 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
