siRNA Details
Gene Symbol: Sh3kbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135727.2 | NP_001129199.1 | 8 | 1231 | ||
NM_001135728.2 | NP_001129200.1 | 3 | 233 | |||
NM_021389.6 | NP_067364.2 | 7 | 796 | |||
GenBank mRNA | AK004636.1 | - | 7 | 796 | ||
AK018032.1 | - | 7 | 812 | |||
AK020782.1 | - | 7 | 773 | |||
AK028299.1 | - | 3 | 234 | |||
AK030839.1 | - | 7 | 797 | |||
AK038540.1 | - | 10 | 1095 | |||
AK049182.1 | - | 8 | 1232 | |||
AK080930.1 | - | 8 | 1096 | |||
AK161735.1 | - | 3 | 314 | |||
AK172022.1 | - | 8 | 1212 | |||
BC034646.1 | - | 7 | 800 | |||
BC052863.1 | - | 7 | 745 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_053360, XM_006256929, XM_006256931, XM_006256930, XM_006256924, and others.
Gene Ontology Categories:
