siRNA Details
Gene Symbol: Zbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021394.2 | NP_067369.2 | 5 | 855 | ||
GenBank mRNA | AF136520.1 | - | 5 | 823 | ||
AK157215.1 | - | 4 | 2196 | |||
AX775969.1 | - | 5 | 809 | |||
AX775971.1 | - | 5 | 809 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
