siRNA Details
Gene Symbol: Pdcd1lg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021396.2 | NP_067371.1 | 3 | 497 | ||
GenBank mRNA | AF142780.2 | - | 3 | 470 | ||
AK089369.1 | - | 3 | 500 | |||
AK141111.1 | - | 2 | 587 | |||
AK155002.1 | - | 3 | 437 | |||
BC104138.1 | - | 3 | 402 | 40045578 | ||
BC104139.1 | - | 3 | 402 | 40045580 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
