siRNA Details
Gene Symbol: Cst10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021405.2 | NP_067380.1 | 1 | 304 | ||
GenBank mRNA | AB036743.1 | - | 1 | 304 | ||
BC048364.1 | - | 1 | 220 | 4920219 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
