siRNA Details
Gene Symbol: Mmp19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164197.1 | NP_001157669.1 | 7 | 1009 | ||
NM_021412.2 | NP_067387.1 | 7 | 1144 | |||
GenBank mRNA | AF153199.1 | - | 7 | 1096 | ||
AF155221.1 | - | 7 | 1124 | |||
AF162446.1 | - | 7 | 1124 | |||
AK079924.1 | - | 7 | 1010 | |||
AK080125.1 | - | 7 | 1005 | |||
AK086808.1 | - | 6 | 884 | |||
AK136517.1 | - | 7 | 998 | |||
AK154863.1 | - | 7 | 1138 | |||
BC112966.2 | - | 7 | 1116 | 40039023 |
