siRNA Details
Gene Symbol: Fam181b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021427.2 | NP_067402.2 | 1 | 257 | ||
GenBank mRNA | AB041569.1 | - | 1 | 217 | ||
AK043957.1 | - | 1 | 272 | |||
AK140685.1 | - | 1 | 209 | |||
BC043329.1 | - | 1 | 206 | 5369836 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
